missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Cinnamtannin A2, ≥85% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 86631-38-1 |
| Product Line | phyproof |
| Linear Formula | C60H50O24 |
| Synonym | Cinnamtannin I; Epicatechin tetramer |
| Type | Reference Substance |
| Quantity | 5 mg |
| Formula Weight | 1155.02 |
| Percent Purity | ≥85% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG