missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Carminic acid, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002371
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002371 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002371 Supplier Merck Emd Millipore Supplier No. PHL8286510MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 1260-17-9
Product Line phyproof
Linear Formula C22H20O13
MDL Number MFCD00167028
Synonym Natural Red 4
Type Reference Substance
Quantity 10 mg
Formula Weight 492.39
Percent Purity ≥95% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG