missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Capsaicin, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002114
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002114 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002114 Supplier Merck Emd Millipore Supplier No. PHL8952025MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 404-86-4
Product Line phyproof
Flash Point 113°C - closed cup
Linear Formula (CH3)2 CHCH=CH(CH2)4 CONHCH2C6H3-4-(OH)-3-(OCH3)
MDL Number MFCD00017259
Melting Point 62°C to 65°C
Synonym 8-Methyl-N-vanillyl-trans-6-nonenamide
Type Reference Substance
Quantity 25 mg
Formula Weight 305.41
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG