missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Bacopaside V, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 620592-16-7 |
| Product Line | phyproof |
| Linear Formula | C41H66O13 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 766.95 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG