missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Bacopaside I, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 382148-47-2 |
| Product Line | phyproof |
| Linear Formula | C46H74O20 S |
| Synonym | Pseudojujubogenin 3-O-[alpha-L-arabinofuranosyl-(1->2)-[6-O-sulfo-beta-D-glucopyranosyl-(1->3)]-alpha-L-arabinopyranoside] |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 979.13 |
| Percent Purity | ≥90% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG