missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Allantoin, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001808
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001808 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001808 Supplier Merck Emd Millipore Supplier No. PHL89518100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 97-59-6
Product Line phyproof
Linear Formula C4H6N4O3
MDL Number MFCD00005260
Melting Point 230°C
Synonym 5-Ureidohydantoin; Glyoxylic(acid) diureide
Type Reference Substance
Quantity 100 mg
Formula Weight 158.12
Percent Purity ≥98% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG