missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 6-Shogaol, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001994
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001994 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001994 Supplier Merck Emd Millipore Supplier No. PHL8979210MG
Only null left

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 555-66-8
Product Line phyproof
Linear Formula C17H24O3
MDL Number MFCD01736094
Synonym [6 ]-Shogaol; 1-(4-Hydroxy-3-methoxyphenyl)-4-decen-3-one
Type Reference Substance
Quantity 10 mg
Formula Weight 276.37
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG