missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ 4-Hydroxyisophthalic acid, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001945
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001945 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001945 Supplier Merck Emd Millipore Supplier No. PHL80863100MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 636-46-4
Product Line phyproof
Linear Formula HOC6H3-1,3-(CO2 H)2
MDL Number MFCD00010391
Synonym 4-Hydroxy 1,3-benzenedicarboxylic acid
Type Reference Substance
Quantity 100 mg
Formula Weight 182.13
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG