missing translation for 'onlineSavingsMsg'
Learn More

Phthalocyanine 93.0+%, TCI America™

Catalog No. P035525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P035525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P035525G Supplier TCI America Supplier No. P035525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 574-93-6
Molecular Formula C32H18N8
Molecular Weight (g/mol) 514.552
MDL Number MFCD00005085
InChI Key IEQIEDJGQAUEQZ-UHFFFAOYSA-N
Synonym phthalocyanine, pigment blue 16, 29h,31h-phthalocyanine, phthalocyanin, heliogen blue g, polymon blue g, lionol blue kw, irgazin blue 3gt, ftalocianina, ci pigment blue 16
PubChem CID 5282330
ChEBI CHEBI:34921
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3

Specifications

Color Blue-Purple
Quantity 25 g
Formula Weight 514.55
Percent Purity ≥93.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Phthalocyanine
EINECSNumber (5)-3302
TSCA Yes