missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic Anhydride, 98+% (1,3-Isobenzofurandione), Reagents
Specifications
Specifications
| Melting Point | 131°C |
| Boiling Point | 284°C |
| Color | White |
| Quantity | 2.5 kg |
| Packaging | Poly Bottle |
| Physical Form | Solid |
| Specific Gravity | 1.527 |
| Grade | Reagent |
| Chemical Name or Material | Phthalic Anhydride, 98+% (1,3-Isobenzofurandione) |