missing translation for 'onlineSavingsMsg'
Learn More

Phthalan, 98%

Numéro de catalogue. p-7080619
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1021706 5 g
AAA1021714 25 g
AAA1021722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1021706 Fournisseur Thermo Scientific Chemicals Code fournisseur A1021706
Il en reste null

CAS: 496-14-0 | C8H8O | 120.151 g/mol

Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4'-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4′-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

Solubility
Low aqueous solubility.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.

Identifiants chimiques

CAS 496-14-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005932
InChI Key SFLGSKRGOWRGBR-UHFFFAOYSA-N
Synonym phthalan, 1,3-dihydroisobenzofuran, isocoumaran, isobenzofuran, 1,3-dihydro, o-xylylene oxide, 2-oxaindan, 1,3-dihydro isobenzofuran, unii-2r6ntq7y6g, ccris 1557, 2r6ntq7y6g
PubChem CID 10327
IUPAC Name 1,3-dihydro-2-benzofuran
SMILES C1C2=CC=CC=C2CO1

Spécifications

Density 1.094
Boiling Point 192°C
Flash Point 63°C (145°F)
Refractive Index 1.546
Quantity 5 g
Beilstein 111932
Solubility Information Low aqueous solubility.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material Phthalan
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 207-815-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only