missing translation for 'onlineSavingsMsg'
Learn More

Phthalamic Acid 98.0+%, TCI America™

Catalog No. P028225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P028225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P028225G Supplier TCI America Supplier No. P028225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-97-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00025476
InChI Key CYMRPDYINXWJFU-UHFFFAOYSA-N
Synonym phthalamic acid, phthalamidic acid, phthalamide acid, phthalic monoamide, phthalic acid monoamide, o-carbamoylbenzoic acid, benzoic acid, 2-aminocarbonyl, benzoic acid, o-carbamoyl, unii-v344h4pf3y, 2-aminocarbonyl benzoic acid
PubChem CID 6957
ChEBI CHEBI:50736
IUPAC Name 2-carbamoylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)N)C(=O)O

Specifications

Melting Point 149°C
Color White
Quantity 25 g
Formula Weight 165.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phthalamic Acid