missing translation for 'onlineSavingsMsg'
Learn More

Hydre d’acide phosphotungstique (cristallin/certifié) ACS, Fisher Chemical™

Numéro de catalogue. p-32788
Modifier l'affichage
Click to view available options
Quantité:
25 g
100 g
500 g
Emballage:
Bouteille en polyéthylène
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
A24825 25 g Bouteille en polyéthylène
A248100 100 g Bouteille en polyéthylène
A248500 500 g Bouteille en polyéthylène
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. A24825 Fournisseur Thermo Fisher Scientific Code fournisseur FLA24825

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Deliquescent

Identifiants chimiques

CAS 12501-23-4
Formule moléculaire H5O41PW12
Poids moléculaire (g/mol) 2880.20
Numéro MDL MFCD00011342
Clé InChI AVFBYUADVDVJQL-UHFFFAOYSA-N
Synonyme phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade
PubChem CID 16212977
Nom de l’IUPAC 12-Tungstophosphate hydrate
SOURIRES O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O

Spécifications

Nom ou substance chimique Phosphotungstic Acid
Couleur Blanc
Point de fusion 89 °C
Métaux lourds (sous forme de Pb) 0,005% max.
Désignation Test Réussi
Emballage Bouteille en polyéthylène
Quantité 25 g
Forme physique Solide
Numéro UN 3260
Niveau Certifié
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Call a physician or Poison Control Center immediately. Call a physician or Poison Control Center immediately. .

NFPA

Health:3
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive
Recommended Storage Réfrigérée