missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Phosphorus Standard, 100 ppm P (306.6 ppm PO43-), Ricca Chemical

Numéro de catalogue. 5856516
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
5856516 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 5856516 Fournisseur Ricca Chemical Company Code fournisseur 5856.516
Il en reste null

Identifiants chimiques

CAS 7778-77-0 , 7732-18-5
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Spécifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1mL = 0.1mg P as Phosphate, 100ppm P (306.6ppm PO43-)
Chemical Name or Material Phosphorus Standard
Afficher plus de résultats Afficher moins de résultats
missing translation for 'shelfLife' 12 months