missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, for HPLC, Solstice

Catalog No. 6001136
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001136 100 mL
6001137 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001136 Supplier Solstice Supplier No. 79606100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for HPLC, 85-90%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Concentration 85 to 90% (T)
Density 1.685g/mL at 25°C
Vapor Pressure 5 mmHg (25°C), 2.2 mmHg (20°C)
Quantity 100 mL
Linear Formula H3PO4
UN Number UN1805
Beilstein 1921286
Formula Weight 98g/mol
Chemical Name or Material Phosphoric acid
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338