missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, Certified, 10.0% (v/v) ±0.2% (v/v), LabChem™

Catalog No. LC186506
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC186506 10 L
LC186501 500 mL
LC186502 1 L
LC186504 4 L
LC186505 20 L
LC18650200 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. LC186506 Supplier LabChem Supplier No. LC186506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Color Colorless
CAS Max % 90, 10
Identification Passes Test
Packaging Cubitainer™
Traceability to NIST Traceable to NIST
Decomposition Information Phosphorus oxides; Thermal decomposition generates: Corrosive vapors
Quantity 10 L
Physical Form Liquid
Linear Formula H3PO4
UN Number UN1805
Solubility Information Soluble in water
Formula Weight 97.1
Percent Purity 10.0% ±0.2% (v/v)
Grade Certified
Chemical Name or Material Phosphoric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature