missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 65% w/w, Reagents

Numéro de catalogue. 01334135
Modifier l'affichage
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
01334135 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01334135 Fournisseur Labchem Inc Code fournisseur CP2239001A
Il en reste null

Phosphoric Acid, 65% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Spécifications

pH 0
Color Colorless
Boiling Point 120°C
Packaging Poly Bottle
Specific Gravity 1.4536
Quantity 1 L
Physical Form Liquid
Percent Purity 65%
Grade Reagent
Chemical Name or Material Phosphoric Acid, 65% w/w