missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 0.25 Molar, Reagents

Numéro de catalogue. 01334423
Modifier l'affichage
Click to view available options
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
01334423 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01334423 Fournisseur Labchem Inc Code fournisseur CP10470020F
Il en reste null

Phosphoric Acid, 0.25 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 7732-18-5 , 7664-38-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name water
SMILES O

Spécifications

pH <2
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0134
Quantity 20 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Phosphoric Acid, 0.25 Molar