missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic Acid Monocyclohexylammonium Salt 98.0+%, TCI America™

Catalog No. P07581G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07581G 1 g
P0758100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07581G Supplier TCI America Supplier No. P07581G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10526-80-4
Molecular Formula C9H18NO6P
Molecular Weight (g/mol) 267.22
MDL Number MFCD00036375
InChI Key VHFCNZDHPABZJO-UHFFFAOYSA-N
Synonym cyclohexanamine 2-phosphonooxy acrylate, phosphoenolpyruvic acid cyclohexylammonium salt, 2-propenoic acid, 2-phosphonooxy-, compd. with cyclohexanamine 1:1, cyclohexylamine; phosphoenolpyruvic acid, phosphoenolpyruvic acid cyclohexylamine salt, phosphoenolpyruvic acid, cyclohexylammonium salt, cyclohexanamine; 2-phosphonooxyprop-2-enoic acid, phospho enol pyruvic acid cyclohexylammonium salt, phosphoenolpyruvic acid monocyclohexylammonium salt
PubChem CID 82702
IUPAC Name 2-(phosphonooxy)prop-2-enoic acid; cyclohexanamine
SMILES NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 267.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phosphoenolpyruvic Acid Monocyclohexylammonium Salt
TSCA Yes
Recommended Storage Refrigerator