missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-VAMP4 (Ser30) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105833
Description
Antibody detects endogenous levels of VAMP4 only when phosphorylated at Ser30.
The vesicle associated membrane proteins (VAMP) or synaptobrevins are calcium binding proteins specific to eukaryotes. VAMPs, along with synaptosomal associated protein of 25 kDa (SNAP-25) and syntaxin, form the core complex of soluble NSF attachment protein receptor (SNARE) proteins that interact with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and alpha-SNAP. These membrane associated proteins play a key role in the regulation of vesicle membrane fusion with the plasma membrane. The Clostridium tetani neurotoxin is a metalloprotease with specificity for VAMP. In Alzheimer's disease, VAMP levels of all isoforms appear to be significantly lowered.Identifiants chimiques
| Phospho-VAMP4 (Ser30) | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| VAMP4 | |
| D1Ertd147e; vamp; VAMP24; VAMP4; VAMP-4; vesicle associated membrane protein 4; vesicle-associated membrane protein 4 | |
| VAMP4 | |
| Primary | |
| Sequential Chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70480, O75379 | |
| 364033, 53330, 8674 | |
| A synthesized peptide derived from human VAMP4(Accession O75379), corresponding to amino acid residues around phosphorylated Ser30. | |
| Antibody |
Spécifications
| Phospho-VAMP4 (Ser30) | |
| Polyclonal | |
| Unconjugated | |
| VAMP4 | |
| D1Ertd147e; vamp; VAMP24; VAMP4; VAMP-4; vesicle associated membrane protein 4; vesicle-associated membrane protein 4 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 364033, 53330, 8674 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70480, O75379 | |
| VAMP4 | |
| A synthesized peptide derived from human VAMP4(Accession O75379), corresponding to amino acid residues around phosphorylated Ser30. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |