Learn More
Invitrogen™ Phospho-Histone H2A.X (Ser139) Monoclonal Antibody (CR55T33), eFluor™ 570, eBioscience™

Description
Description: The CR55T33 monoclonal antibody recognizes phosphorylated serine 139 of human and mouse H2AX. H2AX is a member of the H2A histone family that complex with DNA and other histones to form the repeating nucleosome units characteristic of eukaryotic chromatin. Nucleosomes consist of approximately 147 base pairs of DNA wrapped around an octamer of histones composed of two each of the four histone proteins: H2A, H2B, H3 and H4. After induction of DNA damage such as double-strand breaks by irradiation, genotoxic stresses, replication errors or gene recombination, PI3K-like kinases (e.g., ataxia telangiectasia mutated (ATM), ataxia telangiectasia Rad-3-related (ATR), and DNA-dependent protein kinase (DNA-PK) are activated to phosphorylate serine 139 in H2AX. This early phosphorylation event plays a critical role in recruiting proteins involved in DNA repair. The monoclonal antibody CR55T33 recognizes a single band of approximately 15 kDa on reduced cell lysates from Jurkat cells stimulated with etoposide. Applications Reported: This CR55T33 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This CR55T33 antibody has been tested by immunocytochemistry of methanol-fixed cells and can be used at less than or equal to 1 μg/mL. This CR55T33 antibody has also been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using high p...
Spécifications
Spécifications
| Antigène | Phospho-Histone H2A.X (Ser139) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CR55T33 |
| Concentration | 0.2 mg/mL |
| Conjugué | eFluor 570 |
| Formule | PBS with 0.09% sodium azide; pH 7.2 |
| Gène | H2AX |
| Numéro d’acquisition du gène | P16104, P27661 |
| Alias du gène | AW228881; gamma H2AX; gammaH2ax; H2A histone family member X; H2A histone family, member X; H2A.X; H2A.X variant histone; H2A/X; h2afx; H2ax; H2AX histone; Hist5-2ax; histone 5 protein 2ax; histone H2A.x; Histone H2a/x; Histone H2AX; RGD1566119; similar to H2A histone family, member X; zgc:56329 |
| Afficher plus de résultats |