missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EphA4 (Tyr602) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105119
Description
Antibody detects endogenous levels of EPHA4 only when phosphorylated at Tyr602.
EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.Identifiants chimiques
| Phospho-EphA4 (Tyr602) | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| EPHA4 | |
| 2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1 | |
| EPHA4 | |
| Primary | |
| Sequential chromatography |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P54764, Q03137 | |
| 13838, 2043, 316539 | |
| A synthesized peptide derived from human EPHA4(Accession P54764), corresponding to amino acid residues around phosphorylated Tyr602. | |
| Antibody |
Spécifications
| Phospho-EphA4 (Tyr602) | |
| Polyclonal | |
| Unconjugated | |
| EPHA4 | |
| 2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 13838, 2043, 316539 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P54764, Q03137 | |
| EPHA4 | |
| A synthesized peptide derived from human EPHA4(Accession P54764), corresponding to amino acid residues around phosphorylated Tyr602. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |