missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-eIF4B (Ser406) Recombinant Rabbit Monoclonal Antibody (23GB3905)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553662
Description
Phospho-eIF4B (Ser406) Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
The eukaryotic translation initiation factor 4B (eIF4B) plays a critical role in recruiting the 40S ribosomal subunit to the mRNA. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F.Identifiants chimiques
| Phospho-eIF4B (Ser406) | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Eif4b | |
| 2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
| Eif4b | |
| Primary | |
| Affinity chromatography |
| 23GB3905 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P23588, Q8BGD9 | |
| 1975, 300253, 75705 | |
| A synthetic peptide of human Phospho-eIF4B (Ser406) (399-413AA). | |
| Antibody |
Spécifications
| Phospho-eIF4B (Ser406) | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Eif4b | |
| 2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1975, 300253, 75705 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3905 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P23588, Q8BGD9 | |
| Eif4b | |
| A synthetic peptide of human Phospho-eIF4B (Ser406) (399-413AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |