missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Chk1 (Ser317) Recombinant Rabbit Monoclonal Antibody (Chk1S317-G1)

Numéro de catalogue. PIMA527986
Modifier l'affichage
Click to view available options
Quantité:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIMA527986 200 μL
PIMA537065 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. PIMA527986 Fournisseur Invitrogen™ Code fournisseur MA527986
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

CHEK1 (CHK1) is a serine/threonine protein kinase involved in preventing replication when DNA integrity is compromised. CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.

Spécifications

Antigène Phospho-Chk1 (Ser317)
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone Chk1S317-G1
Concentration 0.5 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol, 0.1% BSA and 0.02% sodium azide; pH 7.4
Gène Chek1
Numéro d’acquisition du gène O14757, O35280
Alias du gène C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W
Symboles de gène Chek1
Espèces hôtes Rabbit
Immunogène A synthetic phospho-peptide corresponding to residues surrounding Ser317 of human phospho Chk1.
Méthode de purification Protein A/G
Quantité 200 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 1111, 12649
Espèces cibles Human, Mouse
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG κ
Afficher plus de résultats Afficher moins de résultats