Learn More
Invitrogen™ Phospho-CaMKII alpha (Thr286) Monoclonal Antibody (22B1)

Description
MA1-047 detects phosphorylated CaM kinase II from human, rat and mouse tissues/cells. This antibody has been shown to detect the phosphorylated alpha subunit CaM kinase II only. MA1-047 does not detect the non-phosphorylated alpha subunit CaM kinase II nor with the ~60 kDa beta subunit in either phosphorylation state. MA1-047 has been successfully used in Western blot, immunofluorescence, immunoprecipitation, immunocytochemistry, immunohistochemistry, and ELISA procedures. By Western blot, this antibody detects an ~50 kDa band representing the alpha subunit of CaM kinase II only after phosphorylation. Immunofluorescence staining of CaM kinase II in rat/mouse hippocampal cells with MA1-047 densely labels cell somas and neuropil areas, while nuclei are only lightly stained. The MA1-047 immunizing peptide is a synthetic peptide corresponding to residues M(281) H R Q E T(p) V D C L K K F N(294) of rat CaM Kinase II protein with a phosphorylated threonine at position 286.
Spécifications
Spécifications
| Antigène | Phospho-CaMKII alpha (Thr286) |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 22B1 |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.05% sodium azide |
| Gène | CAMK2A |
| Numéro d’acquisition du gène | P11275, P11798, Q9UQM7 |
| Alias du gène | alpha CaM kinase II; alpha-CaMKII; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; CaM kinase II alpha subunit; caM kinase II subunit alpha; CaMK II; CAMK2; Camk2a; CAMKA; CaMKII; CaMK-II alpha subunit; caMK-II subunit alpha; CaMKIINalpha; CaM-kinase II alpha chain; EC 2.7.11.17; KIAA0968; mKIAA0968; OTTHUMP00000165787; OTTHUMP00000165788; Phospho-CaMKI; PK2CDD; PKCCD; R74975 |
| Afficher plus de résultats |