missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Standard, 1000 ppm PO43- (326 ppm P), Ricca Chemical

$112.47 - $190.16

Chemical Identifiers

CAS 7732-18-5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5839-4
View Documents
Ricca Chemical Company
RI58394
120 mL Natural Poly Bottle
Each for $112.47
Only null left
 
5839-16
View Documents
Ricca Chemical Company
583916
500 mL Natural Poly Bottle
Each for $190.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5

Specifications

Natural Poly Bottle
1mL = 1mg PO4,1000ppm PO4,326.1ppm Phosphorus (P)
120 mL
7732-18-5
100°C
Phosphate Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'