missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate Standard, 1000 ppm PO43- (326 ppm P), Ricca Chemical
Supplier: Ricca Chemical Company RI58394
Specifications
| Phosphate Standard | |
| 7732-18-5 | |
| 100°C |
| Natural Poly Bottle | |
| 1mL = 1mg PO4,1000ppm PO4,326.1ppm Phosphorus (P) | |
| 120 mL |
Chemical Identifiers
| 7732-18-5 |