missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphatase Inhibitor Cocktail II

Catalog No. AAJ61022AA
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61022AA 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61022AA Supplier Thermo Scientific Chemicals Supplier No. J61022AA

 

The phosphatase inhibitor cocktail improves the yields of intact proteins by adding inhibitors to enzymes that modify proteins present in cell extracts. This product has been optimized and tested for tyrosine protein phosphatases, acid and alkaline phosphatases. It has been tested on cell extracts from animal tissues including bovine liver and human placenta, and cytosolic extract of rabbit muscle. It is a featured product for Kinase Phosphatase Biology research.An inhibitor of L-isozymes of alkaline phosphatase and ser/thr protein phosphatases. It is used with tissue and cell extracts, including extracts containing detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Concentration 100X
Color Colorless
Description Inhibit broad spectrum of serine/theorine and tyrosine phosphatases
Product Type Phosphatase Inhibitor Cocktail
Content And Storage -30°C to -10°C
Form Liquid
Quantity 5 mL
Composition 100mM sodium fluoride, 50mM sodium orthovanadate, 50mM sodium pyrophosphate, and 100mM b-glycerophosphate

RUO – Research Use Only