missing translation for 'onlineSavingsMsg'
Learn More

Phorbol 12-myristate 13-acetate, ≥99%, MP Biomedicals™

Catalog No. ICN19480401
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19480401 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19480401 Supplier MP Biomedicals Inc Supplier No. 0219480401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16561-29-8
Molecular Formula C36H56O8
Molecular Weight (g/mol) 616.84
MDL Number MFCD00036736
InChI Key PHEDXBVPIONUQT-RGYGYFBISA-N
Synonym phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
PubChem CID 27924
ChEBI CHEBI:37537
IUPAC Name (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O

Specifications

Color White to Yellow
Quantity 1 mg
Formula Weight 616.84
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Phorbol 12-myristate 13-acetate