Learn More
Phoenix Diagnostics Blood Gas/ISE Linearity Control Kit
Catalog No. 22024128
Encompass
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
22024128 Each
1 options
Catalog No. 22024128 Supplier Phoenix Diagnostics Supplier No. PH5001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For calibration verification as required by CLIA
  • Protein-based, liquid-stable and ready to use, reducing time and risk
  • Parameters: pH, PCO2, PO2, sodium, potassium, calcium, chloride, lithium, magnesium, creatine, lactate, glucose, BUN
  • Guaranteed minimum stability of 2-3 years eliminates lot-to-lot variability
  • Unique stabilization technology saves time and money, without compromising quality or results
  • Ampules reduce risk of oxygen contamination
Order Info
Kit contains 5 levels x 5 (1mL) ampules per level, for 5 calibration verification runs per level
Specifications
Type Blood Gas/ISE Linearity Control Kit
Includes 5 Levels x 5 (1mL) ampules per level
Level 5
Quantity Each