missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol Reagent, 0.7% w/v, Reagents

Catalog No. 01334407
Change view
Click to view available options
Quantity:
120 mL
55 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334407 120 mL
01334406 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334407 Supplier Labchem Inc Supplier No. CP106700120C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phloroglucinol Reagent, 0.7% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 67-56-1 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 1,3,5-Trihydroxybenzene Dihydrate, 1,3,5-Benzenetriol Dihydrate
IUPAC Name water
SMILES O

Specifications

Color Yellow to Orange
Boiling Point 80°C
Percent Purity 0.7%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.9954
pH <1
Quantity 120 mL
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Phloroglucinol Reagent, 0.7% w/v
Show More Show Less