missing translation for 'onlineSavingsMsg'
Learn More

Phloretin 98.0+%, TCI America™

Catalog No. P19661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19661G 1 g
P19665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19661G Supplier TCI America Supplier No. P19661G

Chemical Identifiers

CAS 60-82-2
Molecular Formula C15H14O5
Molecular Weight (g/mol) 274.272
MDL Number MFCD00002288
InChI Key VGEREEWJJVICBM-UHFFFAOYSA-N
Synonym phloretin, dihydronaringenin, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl propan-1-one, phloretol, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl-1-propanone, unii-s5j5oe47mk, 1-propanone, 3-4-hydroxyphenyl-1-2,4,6-trihydroxyphenyl, ccris 7459, beta-p-hydroxyphenyl phloropropiophenone, 2',4',6'-trihydroxy-3-p-hydroxyphenyl propiophenone
PubChem CID 4788
ChEBI CHEBI:17276
IUPAC Name 3-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one
SMILES C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2O)O)O)O

Specifications

Melting Point 262°C
Quantity 1 g
Formula Weight 274.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phloretin
TSCA No
Recommended Storage Refrigerator