missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ phi29 DNA Polymerase (10 U/μL)

Numéro de catalogue. p-4521346
Change view
Click to view available options
Quantity:
1,000 units
250 units
5,000 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
FEREP0091 250 units
FEREP0092 1,000 units
FEREP0094 5,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. FEREP0091 Fournisseur Thermo Scientific™ Code fournisseur EP0091
Il en reste null

Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification.

Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification. Produced from E. coli cells with a cloned gene 2 of Bacillus subtilis phage phi29, phi29 DNA Polymerase possesses 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.

Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification. Produced from E. coli cells with a cloned gene 2 of Bacillus subtilis phage phi29, phi29 DNA Polymerase possesses 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.

Features of phi29 DNA Polymerase
• Excellent processivity and strand displacement activity—more than 70 kb long DNA stretches can be synthesized
• Highly accurate DNA synthesis
• High yields of amplified DNA even from small amounts of template

Applications
Phi29 DNA Polymerase may be used in a wide variety of applications including:
• Rolling circle amplification (RCA)—generation of periodic DNA nanotemplates
• Multiple displacement amplification (MDA)
• Unbiased amplification of whole genome (WGA) (see figure below)
• Amplification of DNA for SNP and STR detection
• Cell-free amplification of DNA from single cells, pathogenic organisms, or metagenomes
• DNA template preparation for sequencing
• Protein-primed DNA amplification
• Recombination based-cloning
• Cell-free cloning of lethal DNA
• RNA-primed DNA amplification

Notes
1. The addition of pyrophosphatase to the reaction mixture with phi29 DNA Polymerase may enhance DNA synthesis.
2. Because of the enzymes’s 3’→5’ proofreading exonuclease, 3'-modified primers are highly recommended.

Spécifications

Product Type Stand-alone enzyme
For Use With (Application) MDA-WGA, RCA
Polymerase phi29 DNA Polymerase
Quantity 250 units
Content And Storage • phi29 DNA Polymerase (250 units at 10 U/μL)
• 10X Reaction Buffer for phi29 DNA Polymerase
• Store at –15 to –25°C.
Optimal Reaction Temperature 30°C
Shipping Condition Dry Ice
Concentration 10 U/μL
Reaction Time 4 hours

For Research Use Only. Not for use in diagnostic procedures.