missing translation for 'onlineSavingsMsg'
Learn More

Phenytoin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012215
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012215 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012215 Supplier Merck Emd Millipore Supplier No. PHR11391G
Only null left
Pharmaceutical (small molecule)

Specifications

CAS 57-41-0
Physical Form Neat
Linear Formula C15H12N2O2
MDL Number MFCD00005264
Melting Point 293°C to 295°C
Synonym 5,5-Diphenylhydantoin; 5,5-Diphenyl-2,4-imidazolidinedione; Phenytoin
Type Certified Reference Material
Quantity 1 g
Formula Weight 252.27
Grade Certified Reference Material