missing translation for 'onlineSavingsMsg'
Learn More

Phenylurea 98.0+%, TCI America™

Catalog No. P024725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P024725G 25 g
P0247100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P024725G Supplier TCI America Supplier No. P024725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-10-8
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007944
InChI Key LUBJCRLGQSPQNN-UHFFFAOYSA-N
Synonym 1-phenylurea, n-phenylurea, urea, phenyl, phenylcarbamide, monophenylurea, stabilizer vh, stabilisator vh, phenyl-urea, urea, n-phenyl, phenyl urea
PubChem CID 6145
IUPAC Name phenylurea
SMILES C1=CC=C(C=C1)NC(=O)N

Specifications

Melting Point 147°C
Color White-Yellow
Boiling Point 238°C
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Phenylurea
EINECSNumber (9)-1662
RTECSNumber YU0650000
TSCA Yes
Recommended Storage Refrigerator