missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenylmethanesulfonyl fluoride, 99%

Catalog No. AC215740500
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215740500 50 g Glass bottle
AC215740010 1 g Glass bottle
AC215740050 5 g Glass bottle
AC215740100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC215740500 Supplier Thermo Scientific Chemicals Supplier No. 215740500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 329-98-6
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007424
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
PubChem CID 4784
ChEBI CHEBI:8102
IUPAC Name phenylmethanesulfonyl fluoride
SMILES C1=CC=C(C=C1)CS(=O)(=O)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 90.0°C to 94.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2SO2F
Quantity 50 g
Beilstein 11, III, 331
Merck Index 15, 7659
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in xylene and petroleum ether
Formula Weight 174.19
Percent Purity 99%
Physical Form Needles (May Agglomerate)
Chemical Name or Material Phenylmethanesulfonyl fluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minut
EINECSNumber 206-350-2

RUO – Research Use Only