missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine Hydrochloride 98.5+%, TCI America™

Supplier:  TCI America P018425G

 View more versions of this product

Catalog No. P018425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenylhydrazine Hydrochloride
254°C
C6H9ClN2
25 g
phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
[H+].[Cl-].NNC1=CC=CC=C1
144.60
144.60
Crystalline Powder
59-88-1
White-Yellow
MFCD00012924
2811
JOVOSQBPPZZESK-UHFFFAOYSA-N
hydrogen phenylhydrazine chloride
60962
≥98.5% (T)

Chemical Identifiers

59-88-1
144.60
JOVOSQBPPZZESK-UHFFFAOYSA-N
60962
[H+].[Cl-].NNC1=CC=CC=C1
C6H9ClN2
MFCD00012924
phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
hydrogen phenylhydrazine chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0470

RTECSNumber : MV9000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions