missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine Hydrochloride 98.5+%, TCI America™

Catalog No. p-7134475
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
P018425G 25 g
P0184100G 100 g
2 options

Catalog No. P018425G

Supplier: TCI America P018425G

Only null left

Chemical Identifiers

CAS 59-88-1
Molecular Formula C6H9ClN2
Molecular Weight (g/mol) 144.60
MDL Number MFCD00012924
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
PubChem CID 60962
IUPAC Name hydrogen phenylhydrazine chloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1

Specifications

Melting Point 254°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 144.60
Percent Purity ≥98.5% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylhydrazine Hydrochloride
EINECSNumber (3)-0470
RTECSNumber MV9000000
TSCA Yes