accessibility menu, dialog, popup

UserName

CAS RN 59-88-1

IUPAC Name: hydrogen phenylhydrazine chloride
Synonyms: phenylhydrazine
Molecular Weight (g/mol): 144.6
Molecular Formula: C6H9ClN2
InChi Key: JOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=CC=C1
Quantity 
  • (5)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
Grade 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)