missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylhydrazine Hydrochloride, Crystal, Reagent, 98.5-101.5%, Spectrum™ Chemical
P1080, 59-88-1, C6H5NHNH2·HCl
Supplier: Spectrum Chemical Mfg Cor P1080100GM
Description
Spectrum™ Chemical Phenylhydrazine Hydrochloride, Crystal, Reagent , also known as Hydrazinobenzene, is used to prepare indoles by way of the Fischer Indole Synthesis. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.Specifications
| 59-88-1 | |
| 0.0005 | |
| C6H9ClN2 | |
| 100 g | |
| [H+].[Cl-].NNC1=CC=CC=C1 | |
| 144.60 | |
| Reagent |
| 1 | |
| Poly Bottle | |
| MFCD00012924 | |
| JOVOSQBPPZZESK-UHFFFAOYSA-N | |
| hydrogen phenylhydrazine chloride | |
| 98.5 to 101.5% | |
| Crystals or Flakes |
Chemical Identifiers
| 59-88-1 | |
| 144.60 | |
| JOVOSQBPPZZESK-UHFFFAOYSA-N | |
| [H+].[Cl-].NNC1=CC=CC=C1 |
| C6H9ClN2 | |
| MFCD00012924 | |
| hydrogen phenylhydrazine chloride |