missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine Hydrochloride, Crystal, Reagent, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7157982
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605141 25 g Poly Bottle
18605142 100 g Poly Bottle
18605143 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605141 Supplier Spectrum Chemical Mfg Cor Supplier No. P108025GM
Only null left

P1080, 59-88-1, C6H5NHNH2·HCl

Spectrum™ Chemical Phenylhydrazine Hydrochloride, Crystal, Reagent , also known as Hydrazinobenzene, is used to prepare indoles by way of the Fischer Indole Synthesis. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 59-88-1
Molecular Formula C6H9ClN2
Molecular Weight (g/mol) 144.60
MDL Number MFCD00012924
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
IUPAC Name hydrogen phenylhydrazine chloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1

Specifications

CAS Max % 1
Ignition Residue 0.0005
Packaging Poly Bottle
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade Reagent
Physical Form Crystals or Flakes