missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsonic Acid 99.0+%, TCI America™

Catalog No. B002125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B002125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B002125G Supplier TCI America Supplier No. B002125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-05-5
Molecular Formula C6H7AsO3
Molecular Weight (g/mol) 202.041
MDL Number MFCD00002097
InChI Key LVKZSFMYNWRPJX-UHFFFAOYSA-N
Synonym benzenearsonic acid, monophenylarsonic acid, arsonic acid, phenyl, unii-57f9ku116m, kyselina benzenarsonova czech, arsonic acid, as-phenyl, kyselina benzenarsonova, benzenarsonic acid, benzenearsonie acid, acmc-20ajwq
PubChem CID 7365
ChEBI CHEBI:29851
IUPAC Name phenylarsonic acid
SMILES C1=CC=C(C=C1)[As](=O)(O)O

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 25 g
UN Number 3465
Formula Weight 202.04
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylarsonic Acid
RTECSNumber CY3150000
TSCA Yes