missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide Solution, 0.025N (Certified), Fisher Chemical

Supplier:  Thermo Fisher Scientific SP651

Catalog No. SP65-1



Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenylarsine Oxide Solution
0.025N
100°C
C6H5AsO
0.024 to 0.026N
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
O=[As]C1=CC=CC=C1
168.03
CHEBI:75253
0.025N
Amber Glass
1 L
Undesignated
637-03-6 , 1310-73-2 , 7732-18-5
0°C
MFCD00001990
1.0g/cm³
BQVCCPGCDUSGOE-UHFFFAOYSA-N
arsorosobenzene
4778
Certified
Pass Test
6.5
Liquid

Chemical Identifiers

637-03-6
168.03
BQVCCPGCDUSGOE-UHFFFAOYSA-N
4778
arsorosobenzene
C6H5AsO
MFCD00001990
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
CHEBI:75253
O=[As]C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions