missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide Solution, 0.00564N (Certified), Fisher Chemical™

Catalog No. p-213694
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP681 1 L Glass Bottle
SP684 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP681 Supplier Thermo Fisher Scientific Supplier No. SP68 1
Only null left

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 637-03-6 , 1310-73-2 , 7732-18-5
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

Color Undesignated
Boiling Point 100°C
Melting Point 0°C
Concentration 0.00559 to 0.00569N
Density 1.0g/cm³
Grade Certified
Concentration or Composition (by Analyte or Components) 0.00564N
Identification Pass Test
Packaging Glass Bottle
pH 6.5
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Phenylarsine Oxide Solution, 0.00564N
Show More Show Less