missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylarsine Oxide (PAO) Titrant, 0.0375 N, Stabilized, Ricca Chemical
Chemical Identifiers
| CAS | 637-03-6 , 7732-18-5 , 1310-73-2 , 7732-18-5 |
|---|---|
| Molecular Formula | C6H5AsO |
| Molecular Weight (g/mol) | 168.03 |
| MDL Number | MFCD00001990 |
| InChI Key | BQVCCPGCDUSGOE-UHFFFAOYSA-N |
| Synonym | phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 |
| PubChem CID | 4778 |
| ChEBI | CHEBI:75253 |
| IUPAC Name | arsorosobenzene |
| SMILES | O=[As]C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
576016
|
N/A | 500 mL | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
576032
|
N/A | 1 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 637-03-6 , 7732-18-5 , 1310-73-2 , 7732-18-5 | |
| 168.03 | |
| BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
| 4778 | |
| arsorosobenzene |
| C6H5AsO | |
| MFCD00001990 | |
| phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
| CHEBI:75253 | |
| O=[As]C1=CC=CC=C1 |
Specifications
| 637-03-6 , 7732-18-5 , 1310-73-2 , 7732-18-5 | |
| 0.39, 100.0, 0.1 | |
| MFCD00001990 | |
| BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
| arsorosobenzene | |
| 4778 | |
| Stabilized | |
| Phenylarsine Oxide (PAO) Titrant |
| 0.37, 98.0, 0.1 | |
| C6H5AsO | |
| phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
| O=[As]C1=CC=CC=C1 | |
| 168.03 | |
| CHEBI:75253 | |
| 0.0375Normal |