missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide (PAO) Titrant, 0.0375 N, Stabilized, Ricca Chemical

Supplier:  Ricca Chemical Company 576016

Quantity 500 mL
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 576016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

637-03-6 , 7732-18-5 , 1310-73-2 , 7732-18-5
168.03
BQVCCPGCDUSGOE-UHFFFAOYSA-N
4778  
arsorosobenzene
C6H5AsO
MFCD00001990
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
CHEBI:75253
O=[As]C1=CC=CC=C1

Specifications

Phenylarsine Oxide (PAO) Titrant
0.37, 98.0, 0.1
C6H5AsO
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
O=[As]C1=CC=CC=C1
168.03
CHEBI:75253
Stabilized
0.0375Normal
637-03-6 , 7732-18-5 , 1310-73-2 , 7732-18-5
0.39, 100.0, 0.1
MFCD00001990
BQVCCPGCDUSGOE-UHFFFAOYSA-N
arsorosobenzene
4778  
Amber Glass Bottle
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents