missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide (PAO) Titrant, 0.0250 N (N/40), Stabilized, Ricca Chemical

Catalog No. 575516
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
575516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 575516 Supplier Ricca Chemical Company Supplier No. 575516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-03-6 , 7732-18-5 , 1310-73-2 , 7732-18-5
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

Color Colorless to Tan
CAS Min % 0.25, 98.0, 0.1
CAS Max % 0.26, 100.0, 0.1
Grade Laboratory
Packaging Amber Glass Bottle
Treatment(s) Stabilized
pH 6 to 8.5
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.0250Normal
Chemical Name or Material Phenylarsine Oxide (PAO) Titrant
Show More Show Less