missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide (PAO) Titrant, 0.00564 N, Stabilized, 1 mL = 0.2 mg Cl2, Ricca Chemical

Catalog No. 575032
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
575032 1 L Amber Glass Bottle
575016 500 mL Amber Glass Bottle
57501 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 575032 Supplier Ricca Chemical Company Supplier No. 575032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-03-6 , 7732-18-5 , 1310-73-2 , 7732-18-5
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

Color Colorless to Tan
CAS Min % 0.05, 98.0, 0.08
CAS Max % 0.06, 100.0, 0.09
Equivalence 1 mL = 0.2 mg Cl2
Grade Laboratory
Packaging Amber Glass Bottle
Treatment(s) Stabilized
pH 6 to 8.5
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.00564N
Chemical Name or Material Phenylarsine Oxide (PAO) Titrant
Show More Show Less