missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylarsine Oxide (PAO) Titrant, 0.00564 N, Stabilized, 1 mL = 0.2 mg Cl2, Ricca Chemical
Supplier: Ricca Chemical Company RI57501
Specifications
| Phenylarsine Oxide (PAO) Titrant | |
| 637-03-6 , 7732-18-5 , 1310-73-2 , 7732-18-5 | |
| 0.06,100.0,0.09 | |
| MFCD00001990 | |
| BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
| arsorosobenzene | |
| 1 mL = 0.2 mg Cl2 | |
| CHEBI:75253 | |
| Amber Glass Bottle | |
| 6 to 8.5 | |
| Liquid |
| Colorless to Tan | |
| 0.05,98.0,0.08 | |
| C6H5AsO | |
| phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
| O=[As]C1=CC=CC=C1 | |
| 168.03 | |
| 4778 | |
| Laboratory | |
| Stabilized | |
| 4 L | |
| 0.00564N |
Chemical Identifiers
| 637-03-6 | |
| 168.03 | |
| BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
| 4778 | |
| arsorosobenzene |
| C6H5AsO | |
| MFCD00001990 | |
| phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
| CHEBI:75253 | |
| O=[As]C1=CC=CC=C1 |