missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetylene, 98+%

Catalog No. AAA1213936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1213936 500 g
AAA1213914 25 g
AAA1213922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1213936 Supplier Thermo Scientific Chemicals Supplier No. A1213936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 536-74-3 | C8H6 | 102.136 g/mol

Phenylacetylene is involved in the preparation of styrene by reduction using Lindlar catalyst. It is used to study the mechanism of the palladium-catalyzed phenyl acetylene oxidative carbonylation reaction. It is also used in the polymerization process to prepare polyphenylacetylene namely 1,2,4-triphenylbenzene and 1,3,5-triphenylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylacetylene is involved in the preparation of styrene by reduction using Lindlar catalyst. It is used to study the mechanism of the palladium-catalyzed phenyl acetylene oxidative carbonylation reaction. It is also used in the polymerization process to prepare polyphenylacetylene namely 1,2,4-triphenylbenzene and 1,3,5-triphenylbenzene.

Solubility
Immiscible with water.

Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, halogens, alkali metals and oxidizing agents.

Chemical Identifiers

CAS 536-74-3
Molecular Formula C8H6
Molecular Weight (g/mol) 102.136
MDL Number MFCD00008570
InChI Key UEXCJVNBTNXOEH-UHFFFAOYSA-N
Synonym phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
PubChem CID 10821
IUPAC Name ethynylbenzene
SMILES C#CC1=CC=CC=C1

Specifications

Melting Point -45°C
Density 0.93
Boiling Point 142°C to 144°C
Flash Point 27°C (81°F)
Refractive Index 1.548
Quantity 500 g
UN Number UN3295
Beilstein 605461
Sensitivity Light sensitive
Merck Index 14,3861
Solubility Information Immiscible with water.
Formula Weight 102.14
Percent Purity ≥98%
Chemical Name or Material Phenylacetylene
Show More Show Less
Hazard Category H226-H304-H315-H319
Hazard Statement GHS H Statement
H304-H226-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 208-645-1
RTECSNumber DA0780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only